Få mere at vide
Tocris Bioscience™ OXA 06 dihydrochloride
GREENER_CHOICE
Potent ROCK inhibitor
Brand:  Tocris Bioscience™ 5182/10
Encompass
Produktkode 15726645
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
OXA 06 dihydrochloride is a potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.
Tekniske data
Tekniske data
>98%
1825455-91-1
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only