Få mere at vide
Tocris Bioscience™ OXA 06 dihydrochloride
Potent ROCK inhibitor
Brand: Tocris Bioscience™ 5182/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
OXA 06 dihydrochloride is a potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.Tekniske data
| >98% | |
| 1825455-91-1 | |
| 50 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only