Learn More
Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts
Product Code. 12484727 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Buffer drain bottle
Replacement Knob (Pack of 6)
Side Clamps; 1 pair
Unit Size:
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12484727 Side Clamps; 1 pair Each
11520495 Buffer drain bottle Each
11962985 Replacement Knob (Pack of 6) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12484727 Supplier Thermo Scientific™ Supplier No. S3SCL
  • ONLINE EXCLUSIVE PRICE
    484.00 DKK
    475.26 DKK / Pakke med 1
    Spare 8.74 DKK
    1% Slukket
Forventet forsendelse: 10-02-2027
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts.
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S3S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
TRUSTED_SUSTAINABILITY
Specifications
Description Side Clamps; 1 pair
For Use With (Equipment) Owl™ S3S Aluminum-Backed Sequencer System
Product Type Clamps