Learn More
Honeywell™ Overspec Safety Glasses
Catalog Number p-7159664
Click to view available options
Lens Tint:
Clear
Gray
Size:
Medium
Large
OTG protection featuring exceptional peripheral vision/coverage, and a customizable fit.
  • Over-the-glass coverage, allowing workers full protection while wearing prescription eyewear
  • Adjustable and bendable
  • Wire-core temples offer flexibility for a secure, customized fit
  • Ratchetting temples (3 positions) to further customize fit
  • For use in agriculture, manufacturing, and pharmaceutical industries
  • Protection from abrasion, airborne particulates, biohazard, impact
TRUSTED_SUSTAINABILITY
Certifications/Compliance EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C899.1B-D, EC Attestation: C899.3B-D
Lens Tint Clear
For Use With (Application) For use in agriculture, manufacturing, and pharmaceutical industries
Frame Color Blue, Black
Size Medium
Lens Material Polycarbonate
Frame Material Nylon