Learn More
BINDER™ Overheating Alarm
For Use With: ED 56, ED 115, ED 260, ED 720
Brand: BINDER™ 8012-1635
This item is not returnable.
View return policy
Description
- Audible
- Can be switched off
- Adjustable limit on the independent temperature safety device
- Switchable acoustic alarm
Specifications
Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device | |
BINDER Chambers ED 56, ED 115, ED 260, ED 720 | |
Plastic | |
Alarm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Overheating Alarm > For Use With: ED 56, ED 115, ED 260, ED 720
Spot an opportunity for improvement?Share a Content Correction