Learn More
BINDER™ Overheating Alarm
For Use With: ED 56, ED 115, ED 260, ED 720
Brand:  BINDER™ 8012-1635
Product Code. 16474339
  • 6365.00 DKK / Each
Only null left
Estimated Shipment: 04-09-2025
Description
  • Audible
  • Can be switched off
  • Adjustable limit on the independent temperature safety device
  • Switchable acoustic alarm
Specifications
Specifications
Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
BINDER Chambers ED 56, ED 115, ED 260, ED 720
Plastic
Alarm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Overheating Alarm > For Use With: ED 56, ED 115, ED 260, ED 720