Få mere at vide
Invitrogen™ Ovalbumin Monoclonal Antibody (3G2E1D9,3D5F4A8,3G1E1D9)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA515307
Produktkode 11365283
  • 4390.00 DKK / 100µg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
MA5-15307 targets OVA in indirect ELISA, WB applications and shows reactivity with Tag samples. The MA5-15307 immunogen is ovalbumin.
Chicken ovalbumin is the major protein in the white of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
Ovalbumin
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
P01012
OVAL
Ovalbumin.
100 μL
Primary
Chicken
Antibody
IgG1
ELISA, Western Blot
3G2E1D9,3D5F4A8,3G1E1D9, 3D5F4A8, 3G1E1D9
Unconjugated
OVAL
Allergen Gal d II; clade (ovalbumin); egg albumin; Gal d 2; member 14; OVA; OVAL; Ovalbumin; ovalbumin (SERPINB14); plakalbumin; serpin peptidase inhibitor; SERPINB14; unnamed protein product
Mouse
Purified
RUO
396058
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.