Learn More
Vitlab™ Oval Magnetic Stir Bar
Vitlab™ Oval Magnetic Stir Bar with permanent magnet AlNiCo V core.
Brand: Vitlab™ 311697
Additional Details : Weight : 0.01000kg
Description
- Highly suitable for vessels with a round bottom, such as round-bottom flasks
- The angled side surfaces act to produce high turbulence, and thus achieve effective mixing
Specifications
20 mm | |
40 mm | |
Oval |