missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTOP1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06362B
Additional Details : Weight : 0.00970kg
Description
OTOP1 Polyclonal specifically detects OTOP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
OTOP1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC163302, MGC163304, otopetrin 1, otopetrin-1 | |
Produced in rabbits immunized with E. coli-derived Human OTOP1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
133060 | |
Store at 4°C in the dark. | |
IgG |