missing translation for 'onlineSavingsMsg'
Learn More
Otenabant, MedChemExpress Product Code.: 16494228

Otenabant, MedChemExpress

Product Code. 16494228
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494228

Brand: Medchem Express HY10871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Otenabant is a potent and selective cannabinoid receptor CB1 antagonist with Ki of 0.7 nM, exhibits 10,000-fold greater selectivity against human CB2 receptor.

Otenabant is a potent and selective cannabinoid receptor CB1 antagonist with Ki of 0.7 nM, exhibits 10,000-fold greater selectivity against human CB2 receptor.

Specifications

Chemical Name or Material Otenabant
CAS 686344-29-6
Color White
Purity Grade Notes Research
Molecular Formula C25H25Cl2N7O
Quantity 10 mg
Synonym CP-945598
Solubility Information DMSO : 100 mg/mL (195.92 mM; Need ultrasonic)
SMILES ClC1=CC=CC=C1C2=NC3=C(N4CCC(NCC)(C(N)=O)CC4)N=CN=C3N2C5=CC=C(Cl)C=C5
Molecular Weight (g/mol) 510.42
Formula Weight 510.42
Percent Purity 99.33%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Otenabant, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.