Få mere at vide
Tocris Bioscience™ OSU 03012
Beskrivelse
OSU 03012 is a PDPK1 (PDK1) inhibitor; inhibits Akt signaling. Induces apoptosis of PC-3 and medulloblastoma cells, and inhibits growth of a number of tumor cell lines. Sensitizes radiotherapy-induced cell death and enhances cytotoxic effects of chemotherapeutic agents in vitro. Attenuates tumor growth of medulloblastoma xenografts in mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Amino-N-[4-[5-(2-phenanthrenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]phenyl]acetamide |
| CAS | 742112-33-0 |
| Mængde | 10 mg |
| Mål | PDK-1 Inhibitors |
| Molekylær formel | C26H19F3N4O |
| Formel vægt | 460.45 |
| Renhed | >98% |