Få mere at vide
Ostruthin Imperatorin, TRC
Artikelnummer. 30424602
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424602 10 mg
30410104 1 mg
30412724 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30424602 Leverandør Toronto Research Chemicals Leverandørnr. TRCO70390810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148-83-4
Molekylær formel C19H22O3
Molekylvægt (g/mol) 298.38
Synonym Ostruthine
IUPAC navn 6-[(2E)-3,7-dimethylocta-2,6-dienyl]-7-hydroxychromen-2-one
SMIL CC(=CCC\C(=C\Cc1cc2C=CC(=O)Oc2cc1O)\C)C
Tekniske data
Kemisk navn eller materiale Ostruthin Imperatorin
Mængde 10 mg
InChI formel InChI=1S/C19H22O3/c1-13(2)5-4-6-14(3)7-8-15-11-16-9-10-19(21)22-18(16)12-17(15)20/h5,7,9-12,20H,4,6,8H2,1-3H3/b14-7+
Formel vægt 298.1569