Learn More
Osthole, TRC
Product Code. 30134962
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134962 1 g 1g
30006179 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134962 Supplier Toronto Research Chemicals Supplier No. TRCO7035701G
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 484-12-8
Molecular Formula C15 H16 O3
Molecular Weight (g/mol) 244.29
Synonym 2H-1-Benzopyran-2-one, 7-methoxy-8-(3-methyl-2-buten-1-yl)-, 2H-1-Benzopyran-2-one, 7-methoxy-8-(3-methyl-2-butenyl)- (9CI), Coumarin, 7-methoxy-8-(3-methyl-2-butenyl)- (7CI,8CI), Osthole (6CI), 7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-1-benzopyran-2-one, 7-Methoxy-8-(3-methyl-2-butenyl)coumarin, 7-Methoxy-8-isopentenylcoumarin, 8-(3-Methyl-2-butenyl)herniarin, NSC 31868, Osthol, Ostol, Ostole
IUPAC Name 7-methoxy-8-(3-methylbut-2-enyl)chromen-2-one
SMILES COc1ccc2C=CC(=O)Oc2c1CC=C(C)C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H16O3/c1-10(2)4-7-12-13(17-3)8-5-11-6-9-14(16)18-15(11)12/h4-6,8-9H,7H2,1-3H3
Formula Weight 244.1099
Chemical Name or Material Osthole