missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Osimertinib dimesylate, MedChemExpress
Description
Osimertinib dimesylate (AZD-9291 dimesylate) is an irreversible and mutant selective EGFR inhibitor with IC50s of 12 and 1 nM against EGFRL858R and EGFRL858R/T790M, respectively.
Specifications
Specifications
| Kemisk navn eller materiale | Osimertinib dimesylate |
| CAS | 2070014-82-1 |
| Farve | Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C30H41N7O8S2 |
| Mængde | 100 mg |
| Synonym | AZD-9291 dimesylateMereletinib dimesylate |
| Opløselighedsinformation | H2O : 100 mg/mL (144.55 mM; Need ultrasonic) ∣DMSO : 2.63 mg/mL (3.80 mM; Need ultrasonic) |
| SMIL | CS(=O)(O)=O.C=CC(NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C)=O.CS(=O)(O)=O |
| Molekylvægt (g/mol) | 691.82 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?