Få mere at vide
OSI-420-d4, Free Base (Desmethyl Erlotinib-d4), TRC
Artikelnummer. 30410252
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30410252 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410252 Leverandør Toronto Research Chemicals Leverandørnr. TRCO70225210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-O-Desmethylerlotinib phenyl-2,4,5,6-d4
CAS 1216420-11-9
Molekylær formel C21 2H4 H17 N3 O4
Mængde 10 mg
Synonym 2-[7-(2-Methoxyethoxy)-4-[(2,3,4,6-tetradeuterio-5-ethynylphenyl)amino]quinazolin-6-yl]oxyethanol, 2-[[4-[(3-Ethynylphenyl-2,4,5,6-d4)amino]-7-(2-methoxyethoxy)-6-quinazolinyl]oxy]ethanol, CP 473420-d4, Desmethyl erlotinib-d4, OSI-420-d4 (free base), 6-O-Desmethylerlotinib-D4 (phenyl-2,4,5,6-D4), N-(3-Ethynylphenyl-2,4,5,6-d4)-6-(2-hydroxyethoxy)-7-(2-methoxyethoxy)-4-quinazolinamine
InChI formel InChI=1S/C21H21N3O4/c1-3-15-5-4-6-16(11-15)24-21-17-12-19(27-8-7-25)20(28-10-9-26-2)13-18(17)22-14-23-21/h1,4-6,11-14,25H,7-10H2,2H3,(H,22,23,24)/i4D,5D,6D,11D
SMIL [2H]c1c([2H])c(Nc2ncnc3cc(OCCOC)c(OCCO)cc23)c([2H])c(C#C)c1[2H]
IUPAC navn 2-[7-(2-methoxyethoxy)-4-(2,3,4,6-tetradeuterio-5-ethynylanilino)quinazolin-6-yl]oxyethanol
Molekylvægt (g/mol) 383.43
Formel vægt 383.1783