Få mere at vide
ortho-Topolin, TRC
Artikelnummer. 30452069
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452069 25 mg
30451570 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30452069 Leverandør Toronto Research Chemicals Leverandørnr. TRCT54028025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20366-83-0
Molekylær formel C12H11N5O
Molekylvægt (g/mol) 241.25
Synonym 2-[(9H-Purin-6-ylamino)methyl]phenol, 6-(2-Hydroxybenzylamino)purine, 6-(o-Hydroxybenzyl)aminopurine, alpha-(Purin-6-ylamino)-o-cresol, 2-[(1H-Purin-6-ylamino)methyl]-phenol, 6-(2-Hydroxybenzylamino)purine
IUPAC navn 2-[(7H-purin-6-ylamino)methyl]phenol
SMIL Oc1ccccc1CNc2ncnc3nc[nH]c23
Tekniske data
Kemisk navn eller materiale ortho-Topolin
Mængde 25 mg
InChI formel InChI=1S/C12H11N5O/c18-9-4-2-1-3-8(9)5-13-11-10-12(15-6-14-10)17-7-16-11/h1-4,6-7,18H,5H2,(H2,13,14,15,16,17)
Formel vægt 241.0964