Få mere at vide
Orphenadrine Hydrochloride, Mikromol™
Beskrivelse
Orphenadrine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Orphenadrine Hydrochloride |
| CAS | 341-69-5 |
| Analyt- eller komponentnavne | Orphenadrine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H23 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Orphenadrine Hydrochloride, N,N-Dimethyl-2-[(2-methylphenyl)phenylmethoxy]ethanamine hydrochloride, o-Methyldiphenhydramine hydrochloride, o-Monomethyldiphenhydramine hydrochloride, 2-(Phenyl-o-tolylmethoxy)ethyldimethylamine hydrochloride, Phenyl-o-tolylmethyl dimethylaminoethyl ether hydrochloride, Disipal, Mephenamin, N,N-Dimethyl-2-(o-methyl-alpha-phenylbenzyloxy)ethylamine hydrochloride, beta-Dimethylaminoethyl 2-methylbenzhydryl ether hydrochloride |
| InChI formel | InChI=1S/C18H23NO.ClH/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;/h4-12,18H,13-14H2,1-3H3;1H |
| SMIL | Cl.CN(C)CCOC(c1ccccc1)c2ccccc2C |
| IUPAC navn | N,N-dimethyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride |
| Vis mere |