missing translation for 'onlineSavingsMsg'
Få mere at vide

Orphenadrine EP E (as Hydrochloride), Mikromol™

Artikelnummer. 30462083
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462083 4 x 25 mg
30050790 25 mg
30051783 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30462083 Leverandør Mikromol Leverandørnr. MM0801.0500254AMP

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Orphenadrine EP E (as Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (RS)-N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine Hydrochloride (meta-Methylbenzyl Isomer Hydrochloride)
CAS 17630-53-4
Analyt- eller komponentnavne (RS)-N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine Hydrochloride (meta-Methylbenzyl Isomer Hydrochloride)
Forsendelsestilstand Room Temperature
Molekylær formel C18 H23 N O . Cl H
Certificeringer/compliance ISO 9001
Synonym Orphenadrine Hydrochloride Imp. E (EP), Orphenadrine Imp. E (EP), Orphenadrine USP Related Compound A (AS), Orphenadrine USP RC A (AS), (RS)-N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. E (EP) as Hydrochloride, meta-Methylbenzyl Isomer Hydrochloride of Orphenadrine, Ethanamine, N,N-dimethyl-2-[(3-methylphenyl)phenylmethoxy]-, hydrochloride (1:1), Ethanamine, N,N-dimethyl-2-[(3-methylphenyl)phenylmethoxy]-, hydrochloride (9CI), Ethylamine, N,N-dimethyl-2-[(m-methyl-alpha-phenylbenzyl)oxy]-, hydrochloride (8CI), Orphenandrine, Orphenadrine Related Compound E as Hydrochloride, Orphenadrine Hydrochloride Impurity E as Hydrochloride, Orphenadrine Impurity E as Hydrochloride
InChI formel InChI=1S/C18H23NO.ClH/c1-15-8-7-11-17(14-15)18(20-13-12-19(2)3)16-9-5-4-6-10-16;/h4-11,14,18H,12-13H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.CN(C)CCOC(c1ccccc1)c2cccc(C)c2
IUPAC navn N,N-dimethyl-2-[(3-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
Molekylvægt (g/mol) 305.84
Mængde 4 x 25 mg
Formel vægt 305.1546 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.