Få mere at vide
Orphenadrine EP E (as Hydrochloride), Mikromol™
Beskrivelse
Orphenadrine EP E (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (RS)-N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine Hydrochloride (meta-Methylbenzyl Isomer Hydrochloride) |
| CAS | 17630-53-4 |
| Analyt- eller komponentnavne | (RS)-N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine Hydrochloride (meta-Methylbenzyl Isomer Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H23 N O . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Orphenadrine Hydrochloride Imp. E (EP), Orphenadrine Imp. E (EP), Orphenadrine USP Related Compound A (AS), Orphenadrine USP RC A (AS), (RS)-N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. E (EP) as Hydrochloride, meta-Methylbenzyl Isomer Hydrochloride of Orphenadrine, Ethanamine, N,N-dimethyl-2-[(3-methylphenyl)phenylmethoxy]-, hydrochloride (1:1), Ethanamine, N,N-dimethyl-2-[(3-methylphenyl)phenylmethoxy]-, hydrochloride (9CI), Ethylamine, N,N-dimethyl-2-[(m-methyl-alpha-phenylbenzyl)oxy]-, hydrochloride (8CI), Orphenandrine, Orphenadrine Related Compound E as Hydrochloride, Orphenadrine Hydrochloride Impurity E as Hydrochloride, Orphenadrine Impurity E as Hydrochloride |
| InChI formel | InChI=1S/C18H23NO.ClH/c1-15-8-7-11-17(14-15)18(20-13-12-19(2)3)16-9-5-4-6-10-16;/h4-11,14,18H,12-13H2,1-3H3;1H |
| SMIL | Cl.CN(C)CCOC(c1ccccc1)c2cccc(C)c2 |
| IUPAC navn | N,N-dimethyl-2-[(3-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride |
| Vis mere |