missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ornithine Carbamoyltransferase Antibody (CL4045) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59038AF700
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ornithine Carbamoyltransferase Monoclonal antibody specifically detects Ornithine Carbamoyltransferase in Cynomolgus Monkey,Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Tekniske data
| Ornithine Carbamoyltransferase | |
| Monoclonal | |
| Alexa Fluor 700 | |
| EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase | |
| This antibody was developed using a recombinant protein derived from P00480, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| metabolism | |
| 5009 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL4045 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey, Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion