Få mere at vide
Ornidazole Isomer (Impurity), TRC
Artikelnummer. 30422925
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422925 2.5 g
30422775 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30422925 Leverandør Toronto Research Chemicals Leverandørnr. TRCO6885052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14419-11-5
Molekylær formel C7 H10 Cl N3 O3
Molekylvægt (g/mol) 219.63
Synonym 1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole
IUPAC navn 1-chloro-3-(2-methyl-4-nitroimidazol-1-yl)propan-2-ol
SMIL Cc1nc(cn1CC(O)CCl)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 1-(3-Chloro-2-hydroxypropyl)-2-methyl-4-nitroimidazole
Mængde 2.5 g
InChI formel InChI=1S/C7H10ClN3O3/c1-5-9-7(11(13)14)4-10(5)3-6(12)2-8/h4,6,12H,2-3H2,1H3
Formel vægt 219.0411