missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ Orlistat
GREENER_CHOICE

Artikelnummer. 18074962
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18074962 50 mg
18015912 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18074962 Leverandør Tocris Bioscience™ Leverandørnr. 3540/50

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Pancreatic, gastric and carboxylester lipase inhibitor; antiobesity and antihypercholesterolemic activity

Hypolipemic pancreatic, gastric and carboxylester lipase inhibitor. Exhibits no activity at phospholipase A2, liver esterase, trypsin and chymotrypsin. Inhibits the thioesterase domain of fatty acid synthase, leading to cell cycle arrest at the G1/S boundary in vitro. Prevents the absorption of approximately one third of fat from food and exhibits progastrokinetic, antiobesity and antihypercholesterolemic activity in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96829-58-2
Molekylær formel C29H53NO5

Specifications

Kemisk navn eller materiale Orlistat
Mængde 50 mg
Formel vægt 495.73
Renhed >99%
Molekylvægt (g/mol) 495.75
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.