missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR5D13 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06140AF750
This item is not returnable.
View return policy
Description
OR5D13 Polyclonal specifically detects OR5D13 in Human samples. It is validated for Western Blot.
Specifications
OR5D13 | |
Polyclonal | |
Western Blot | |
olfactory receptor 5D13, Olfactory receptor OR11-142, Olfactory receptor OR11-148, olfactory receptor, family 5, subfamily D, member 13 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human OR5D13. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
390142 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction