Learn More
Corning™ LSE™ Optional Head
Product Code. 11981981 Shop All Corning Products
Change view
Click to view available options
Holds:
12 x 1.5 to 2 mL Tubes
2 x 50 mL Tubes
4 x 15 mL Tubes
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11981981 2 x 50 mL Tubes Each
11961981 12 x 1.5 to 2 mL Tubes Each
11971981 4 x 15 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11981981 Supplier Corning™ Supplier No. 480107
  • ONLINE EXCLUSIVE PRICE
    750.00 DKK
    700.53 DKK / Pakke med 1
    Spare 49.47 DKK
    6% Slukket
Forventet forsendelse: 09-04-2027
For use with various sizes of tubes.
This is an optional head for tubes, held horizontally, for use with Corning™ LSE™ Vortex Mixer.
TRUSTED_SUSTAINABILITY
Specifications
Holds 2 x 50 mL Tubes
Product Type Optional Head
For Use With (Equipment) Corning™ LSE™ Vortex Mixer