Få mere at vide
SHP Steriltechnik AG Option V Laboklav 55-195, Vacuum
Beskrivelse
- Permanent or fractional pre-vacuum for optimal air removal
- Reliable penetration of steam into sterilisation items that are difficult to vent
- Fractional pre-vacuum ensures reproducible sterilisation, DIN 58951-2
- Drying vacuum for efficient post-drying of packaged items
- Condensate is reliably removed during the drying phase
- Avoids the need for additional transfer to a drying cabinet
- Suitable for solid, moist or packaged items
Tekniske data
Tekniske data
| Til brug med (udstyr) | Autoclaves, Laboklav 55-195 |
| Model- | Option V Laboklav 55-195 |
| Produkttype | Autoclave Vacuum |