Få mere at vide
Thermo Scientific Chemicals Optical Windows
Artikelnummer. 15447035
Diameter (metrisk):
25,4 mm
50,8 mm
Tykkelse (metrisk):
2 mm
Materiale:
Cadmium Telluride
Galliumarsenid
Kvarts
Safir
Oversigt:
Overfladefinish: 40/20
Overfladefinish: 60/40
Pakningsstørrelse:
Hver
Artikelnummer. 15447035
Brand: Thermo Scientific Chemicals 045636.KS
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Available in different materials and dimensions
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Tekniske data
Oversigt Overfladefinish: 60/40
Materiale Galliumarsenid
Diameter (metrisk) 50,8 mm
Diameter (engelsk) 2 in.
Produkttype Optical Window
Tykkelse (metrisk) 2 mm