missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sherwood Scientific™ Optical Gelatin Filter
540.00 DKK - 842.00 DKK
Product Code | Brand | Spectral Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Spectral Range | Price | Quantity & Availability | |||||
12772675
|
Sherwood Scientific™
25216001 |
470 nm |
770.00 DKK
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12782675
|
Sherwood Scientific™
25217001 |
490 nm |
770.00 DKK
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12762675
|
Sherwood Scientific™
25215001 |
430 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12752685
|
Sherwood Scientific™
25231001 |
410 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12792675
|
Sherwood Scientific™
25218001 |
520 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12702685
|
Sherwood Scientific™
25219001 |
540 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12712685
|
Sherwood Scientific™
25220001 |
580 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12732685
|
Sherwood Scientific™
25222001 |
710 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12722685
|
Sherwood Scientific™
25221001 |
600 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Optical Gelatin Filter | |
1 Ea. |