Få mere at vide
Opipramol Dihydrochloride, LoGiCal
Artikelnummer. 30459416
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459416 10 mg
1 options
Artikelnummer. 30459416 Leverandør LoGiCal Leverandørnr. LGCFOR0422.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Opipramol Dihydrochloride
CAS 909-39-7
Analyt- eller komponentnavne Opipramol Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C23 H29 N3 O . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym 1-Piperazineethanol, 4-[3-(5H-dibenz[b,f]azepin-5-yl)propyl]-, hydrochloride (1:2), 1-Piperazineethanol, 4-[3-(5H-dibenz[b,f]azepin-5-yl)propyl]-, dihydrochloride (7CI,8CI,9CI), 5H-Dibenz[b,f]azepine, 1-piperazineethanol deriv., 4-[3-(5H-Dibenz[b,f]azepin-5-yl)propyl]-1-piperazineethanol dihydrochloride, Dinsidon, G 33040, Insidon, Insidone, Nisidana, Opipramol hydrochloride, Pramolan, 5-[gamma-(beta-Hydroxyethylpiperazino)propyl]-5H-dibenzo[b,f]azepine dihydrochloride
InChI formel InChI=1S/C23H29N3O.2ClH/c27-19-18-25-16-14-24(15-17-25)12-5-13-26-22-8-3-1-6-20(22)10-11-21-7-2-4-9-23(21)26;;/h1-4,6-11,27H,5,12-19H2;2*1H
Løsningstype Neat
SMIL Cl.Cl.OCCN1CCN(CCCN2c3ccccc3C=Cc4ccccc24)CC1
IUPAC navn 2-[4-(3-benzo[b][1]benzazepin-11-ylpropyl)piperazin-1-yl]ethanol;dihydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 436.42
Mængde 10 mg
Formel vægt 435.1844 g/mol