Få mere at vide
Opipramol-d4, TRC
Artikelnummer. 30427784
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30427784 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427784 Leverandør Toronto Research Chemicals Leverandørnr. TRCO66960210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Opipramol-d4
CAS 1215716-70-3
Molekylær formel C23H25D4N3O
Mængde 10 mg
Synonym 4-[3-(5H-Dibenz[b,f]azepin-5-yl)propyl]-1-piperazineethanol-d4, N-[3-[4-(2-Hydroxyethyl)piperazino]propyl]iminostilbene-d4
InChI formel InChI=1S/C23H29N3O/c27-19-18-25-16-14-24(15-17-25)12-5-13-26-22-8-3-1-6-20(22)10-11-21-7-2-4-9-23(21)26/h1-4,6-11,27H,5,12-19H2/i18D2,19D2
SMIL OC([2H])([2H])C([2H])([2H])N(CC1)CCN1CCCN2C(C=CC=C3)=C3C=CC4=CC=CC=C42
Molekylvægt (g/mol) 367.52
Formel vægt 367.2562