missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jaytec Glass™ Open Top Borosilicate Glass Dropping Funnels
Open Top Borosilicate Glass Dropping Funnels for easy refilling in situ. The stopcock permits carefully controlled flow of reagents.
Brand: Jaytec Glass™ FPM-210-070X
Additional Details : Weight : 0.01000kg
Description
- Open top
- Cylindrical shape
- With interchangeable PTFE stopcock
Specifications
Dropping funnel | |
100 mL | |
Borosilicate Glass | |
No | |
Yes |
40 mm | |
8 mm | |
272 mm | |
PTFE stopcock key |