Learn More
Kolmi Hopen™ Op-R™ Type II Medical Masks with Earloops
Pleated medical mask with anti-fog nose clip for patients and healthcare workers.
Brand: Kolmi Hopen™ M13201-30
Additional Details : Weight : 3.70500kg
Description
- High filtration media (polypropylene)
- Two wide and comfortable elastic bands
- Non-woven, soft bias binding
- Integrated anti-fog nose clip
- Filter without fiberglass
- Produced in ISO 8 clean room (particulate and microbiological control)
Specifications
Green | |
Polypropylene | |
CE, ISO 13485, (EU) 2017/745, EN 14683, EN ISO 14971 | |
Medical |
Earloops | |
Pleated | |
Yes | |
60 Pk. |