Learn More
Kolmi Hopen™ Op-Air™ PRO Oxygen Type IIR FFP2 Respiratory Mask
This respiratory pouch mask is ideal for protection against infectious diseases and in dusty environments.
Brand:  Kolmi Hopen™ M52010S-OR
Additional Details : Weight : 4.56000kg
 View more versions of this product
Product Code. 17730219
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Filtering half mask with FFP2 NR D efficiency class
  • Developed to protect against solid and liquid aerosols and particles
  • Protects environment against droplets emitted by wearer and protects wearer from potential body fluid splashes
  • Knitted head loops are ultrasonically welded for optimal comfort
  • Fits perfectly to face to prevent exposure
  • Made in France
Specifications
Specifications
(EU) 2017/745, EN 14683:2019+AC:2019
Pouch/Duckbill
Polypropylene
Yes
50 Pk.
Orange
Single-use
Medium
Polybags