Få mere at vide
Ondansetron-d3 Hydrochloride Salt, TRC
Artikelnummer. 30412262
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412262 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412262 Leverandør Toronto Research Chemicals Leverandørnr. TRCO6550021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ondansetron-d3 Hydrochloride Salt
CAS 1346605-02-4
Molekylær formel C18H16D3N3O • HCl
Mængde 1 mg
Synonym 1,2,3,9-Tetrahydro-9-(methyl-d3)-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one Hydrochloride, GR 38032-d3, GR 38032X-d3, Zofran-d3, Zophren-d3, Zudan-d3
InChI formel InChI=1S/C18H19N3O.ClH/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2;/h3-6,9-10,13H,7-8,11H2,1-2H3;1H/i2D3;
SMIL O=C1C(CN2C(C)=NC=C2)CCC3=C1C(C=CC=C4)=C4N3C([2H])([2H])[2H].Cl
IUPAC navn 3-[(2-methylimidazol-1-yl)methyl]-9-(trideuteriomethyl)-2,3-dihydro-1H-carbazol-4-one;hydrochloride
Molekylvægt (g/mol) 296.393646
Formel vægt 332.1483