missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Oncostatin M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120974
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Positive test controls include: HepG2, NIH/3T3, Mouse uterus, Mouse heart. The target is usually found in the following locations: Secreted.
Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi's sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi's sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF.
Tekniske data
| Oncostatin M | |
| Polyclonal | |
| Unconjugated | |
| OSM | |
| MGC20461; OncoM; oncostatin M; oncostatin-M; OSM | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18413, 289747, 5008 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13725, P53347, Q65Z15 | |
| OSM | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion