Learn More
Omnifit™ HiT Chromatography Columns
Omnifit™ HiT Chromatography Columns withstand temperatures up to 302°F (150°C)—use with heating block or jacket
Brand:  Kinesis 006HCC-10-10-AF
Additional Details : Weight : 0.34000kg
 View more versions of this product
Product Code. 15855554
  • 3803.68 DKK / Each
Estimated Shipment
23-11-2018
Add to basket
Includes: Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end and a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits).
Description
Description
  • Adjustable endpiece design for quick and easy height adjustment
  • Ideal for both in batch production or within a flow chemistry systems
  • Reusable columns let you pack your own media specific to your application
Specifications
Specifications
Omnifit™ HiT™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 100mm; 1/ea
Liquid Chromatography (LC)
(1) fixed and (1) adjustable
10 μm and 30 μm
100mm
150°C
4-150°C
10mm
600 psi
0.2 to 5.5mL
Borosilicate glass, PEEK, FKM/FPM, PTFE
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end and a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits).
Videos