Få mere at vide
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces Artikelnummer.: 15845484
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces
Artikelnummer. 15845484
400 mm, 25 mm, 103.1 to 181.6 mL, Hver
Encompass_Preferred
Længde (metrisk):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (metrisk) Indvendig:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Kolonnekapacitet:
0.1 to 0.7 mL
0.1 to 2.4 mL
0.1 to 4.1 mL
0.2 to 5.5 mL
0.2 to 9.4 mL
0.4 to 12.4 mL
0.4 to 21.2 mL
1.2 to 34.4 mL
1.2 to 58.9 mL
10.6 to 38.9 mL
103.1 to 181.6 mL
11 to 23.6 mL
120.2 to 440.7 mL
134.7 to 288.6 mL
152.2 to 230.7 mL
16.5 to 29.1 mL
2.1 to 7.5 mL
2.3 to 115.5 mL
202.0 to 356.0 mL
24.3 to 36.9 mL
24.7 to 53.0 mL
27.7 to 211.7 mL
280.4 to 600.9 mL
29.5 to 108.0 mL
37.1 to 65.4 mL
4.7 to 17.3 mL
4.8 to 10.3 mL
420.6 to 741.1 mL
54.8 to 83.1 mL
620.9 to 941.4 mL
68.7 to 147.3 mL
7.2 to 12.7 mL
Pakningsstørrelse:
Hver
Artikelnummer. 15845484
Brand: Diba 006EZS2540AA
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Tekniske data
Temperaturområde (metrisk). 4 to 40°C
Diameter (metrisk) Indvendig 25 mm
Type Liquid Chromatography Column
Maks. Tryk 150 psi
Porestørrelse 10 and 30 μm
Kolonnekapacitet 103.1 to 181.6 mL
Længde (metrisk) 400 mm
Materiale Borosilicate Glass, PTFE, Chemraz™
Maks. Temperatur 40°C
Inkluderer Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting.
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces