Få mere at vide
Diba Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces
Længde (metrisk):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (metrisk) Indvendig:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Kolonnekapacitet:
0.7 mL
2.4 mL
4.1 mL
5.5 mL
7.5 mL
9.4 mL
10.3 mL
12.4 mL
12.7 mL
17.3 mL
21.2 mL
23.6 mL
29.1 mL
34.4 mL
36.9 mL
38.9 mL
53 mL
58.9 mL
65.4 mL
83.1 mL
108 mL
115.5 mL
147.3 mL
181.6 mL
211.7 mL
230.7 mL
288.6 mL
356 mL
440.7 mL
600.9 mL
741.1 mL
941.4 mL
Pakningsstørrelse:
Hver
Beskrivelse
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Tekniske data
Tekniske data
| Temperaturområde (metrisk). | 4 to 40°C |
| Diameter (metrisk) Indvendig | 10 mm |
| Type | Liquid Chromatography Column |
| Maks. Tryk | 600 psi |
| Porestørrelse | 10 and 30 μm |
| Kolonnekapacitet | 9.4 mL |
| Længde (metrisk) | 150 mm |
| Materiale | Borosilicate Glass, PTFE, Polyethylene, FKM/FPM |
| Maks. Temperatur | 40°C |
| Inkluderer | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF Polypropylene fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.