Få mere at vide
- Home
- Products
- Kromatografisøjler og patroner
- LC-kolonner med lavt til medium tryk
- Diba Omnifit™ EZ Chromatography Columns with Two Adjustable Endpieces
Diba Omnifit™ EZ Chromatography Columns with Two Adjustable Endpieces
Artikelnummer. 15865374
Skift visning
Længde (metrisk):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (metrisk) Indvendig:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Kolonnekapacitet:
0.1 to 0.7 mL
0.2 to 5.5 mL
0.2 to 9.4 mL
0.4 to 12.4 mL
0.4 to 21.2 mL
1.2 to 34.4 mL
1.2 to 58.9 mL
10.6 to 38.9 mL
103.1 to 181.6 mL
11 to 23.6 mL
120.2 to 440.7 mL
134.7 to 288.6 mL
152.2 to 230.7 mL
16.5 to 29.1 mL
2.3 to 115.5 mL
202.0 to 356.0 mL
24.3 to 36.9 mL
24.7 to 53.0 mL
280.4 to 600.9 mL
29.5 to 108.0 mL
37.1 to 65.4 mL
4.7 to 17.3 mL
4.8 to 10.3 mL
420.6 to 741.1 mL
54.8 to 83.1 mL
57.7 to 211.7 mL
620.9 to 941.4 mL
68.7 to 147.3 mL
7.2 to 12.7 mL
Pakningsstørrelse:
Hver
29 product options available for selection
| Artikelnummer. | Længde (metrisk) | Diameter (metrisk) Indvendig | Kolonnekapacitet | Pakningsstørrelse |
|---|---|---|---|---|
| 15865374 | 100 mm | 25 mm | 1.2 to 34.4 mL | Hver |
| 15825324 | 50 mm | 6.6 mm | 0.1 to 0.7 mL | Hver |
| 15845334 | 330 mm | 6.6 mm | 4.8 to 10.3 mL | Hver |
| 15875334 | 400 mm | 6.6 mm | 7.2 to 12.7 mL | Hver |
| 15805344 | 100 mm | 10 mm | 0.2 to 5.5 mL | Hver |
| 15835344 | 150 mm | 10 mm | 0.2 to 9.4 mL | Hver |
| 15865344 | 250 mm | 10 mm | 4.7 to 17.3 mL | Hver |
| 15895344 | 330 mm | 10 mm | 11 to 23.6 mL | Hver |
| 15825354 | 400 mm | 10 mm | 16.5 to 29.1 mL | Hver |
| 15855354 | 500 mm | 10 mm | 24.3 to 36.9 mL | Hver |
| 15885354 | 100 mm | 15 mm | 0.4 to 12.4 mL | Hver |
| 15815364 | 150 mm | 15 mm | 0.4 to 21.2 mL | Hver |
| 15845364 | 250 mm | 15 mm | 10.6 to 38.9 mL | Hver |
| 15875364 | 330 mm | 15 mm | 24.7 to 53.0 mL | Hver |
| 15805374 | 400 mm | 15 mm | 37.1 to 65.4 mL | Hver |
| 15835374 | 500 mm | 15 mm | 54.8 to 83.1 mL | Hver |
| 15895374 | 150 mm | 25 mm | 1.2 to 58.9 mL | Hver |
| 15825384 | 250 mm | 25 mm | 29.5 to 108.0 mL | Hver |
| 15855384 | 330 mm | 25 mm | 68.7 to 147.3 mL | Hver |
| 15885384 | 400 mm | 25 mm | 103.1 to 181.6 mL | Hver |
| 15815394 | 500 mm | 25 mm | 152.2 to 230.7 mL | Hver |
| 15845394 | 150 mm | 35 mm | 2.3 to 115.5 mL | Hver |
| 15875394 | 250 mm | 35 mm | 57.7 to 211.7 mL | Hver |
| 15805404 | 330 mm | 35 mm | 134.7 to 288.6 mL | Hver |
| 15835404 | 400 mm | 35 mm | 202.0 to 356.0 mL | Hver |
| 15865404 | 250 mm | 50 mm | 120.2 to 440.7 mL | Hver |
| 15895404 | 330 mm | 50 mm | 280.4 to 600.9 mL | Hver |
| 15825414 | 400 mm | 50 mm | 420.6 to 741.1 mL | Hver |
| 15855414 | 500 mm | 50 mm | 620.9 to 941.4 mL | Hver |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
Denne vare kan ikke returneres.
Se returpolicy
Artikelnummer.
15865374
Leverandør
Diba
Leverandørnr.
006EZ2510AA
Log ind eller registrer dig for at se din pris og tilgængelighed.
På lager
Log ind for at se lagerbeholdningen.
Log ind for at se lagerbeholdningen.
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Indeholder
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.
- PEEK™ column threads offer outstanding durability
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Tekniske data
Tekniske data
| Maks. Tryk | 150 psi |
| Porestørrelse | 10 and 30 μm |
| Kolonnekapacitet | 1.2 to 34.4 mL |
| Længde (metrisk) | 100 mm |
| Materiale | Borosilicate Glass, PTFE, Polyethylene, FKM/FPM |
| Maks. Temperatur | 40°C |
| Inkluderer | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
| Temperaturområde (metrisk). | 4 to 40°C |
| Type | Liquid Chromatography Column |
| Diameter (metrisk) Indvendig | 25 mm |