Learn More
Omnifit™ EZ Chromatography Columns with Two Adjustable Endpieces
Omnifit™ EZ Chromatography Columns with Two Adjustable Endpieces are ergonomically designed endpieces provide simple and quick height adjustment
Brand:  Kinesis 006EZ-06-10-AA
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15855324
  • 3569.38 DKK / Each
Estimated Shipment
28-02-2019
Add to basket
Includes: Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal® 10-32 PEEK conical port fitting.
Description
Description
  • PEEK™ column threads offer outstanding durability
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Specifications
Omnifit™ EZ Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 100mm; 1/ea
Liquid Chromatography (LC)
900 psi
0.1 to 2.4mL
Borosilicate glass, PTFE, PE, FKM/FPM
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.
4-40°C
6.6mm
10 μm and 30 μm
100mm
40°C
Videos