Få mere at vide
Omethoate-d3, TRC
Artikelnummer. 30420801
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30420801 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420801 Leverandør Toronto Research Chemicals Leverandørnr. TRCO6355021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Omethoate-d3
CAS 1189510-77-7
Molekylær formel C5H9D3NO4PS
Mængde 1 mg
InChI formel InChI=1S/C5H12NO4PS/c1-6-5(7)4-12-11(8,9-2)10-3/h4H2,1-3H3,(H,6,7)/i1D3
SMIL [2H]C([2H])([2H])NC(=O)CSP(=O)(OC)OC
Molekylvægt (g/mol) 216.21
Formel vægt 216.0413