missing translation for 'onlineSavingsMsg'
Learn More

omega-Aminocaprylic acid, 99+%, Thermo Scientific Chemicals

Catalog Number p-54407
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1002-57-9
C8H17NO2
159.23
MFCD00008245
UQXNEWQGGVUVQA-UHFFFAOYSA-N
8-aminocaprylic acid, octanoic acid, 8-amino, omega-aminocaprylic acid, 8-amino caprylic acid, 8-amino-octanoic acid, .omega.-aminocaprylic acid, 8-amino capylic acid, 8ac, 8-aminocapylic acid, pubchem13165
66085
8-aminooctanoic acid
NCCCCCCCC(O)=O
This item is not returnable. View return policy
1002-57-9
C8H17NO2
159.23
MFCD00008245
UQXNEWQGGVUVQA-UHFFFAOYSA-N
8-aminocaprylic acid, octanoic acid, 8-amino, omega-aminocaprylic acid, 8-amino caprylic acid, 8-amino-octanoic acid, .omega.-aminocaprylic acid, 8-amino capylic acid, 8ac, 8-aminocapylic acid, pubchem13165
66085
8-aminooctanoic acid
NCCCCCCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 194°C to 196°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2N(CH2)7CO2H
Quantity 1 g
Formula Weight 159.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material ω-Aminocaprylic acid, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • w-Aminocaprylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'