missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ Oltipraz

Description
Oltipraz is a Nrf2 activator; activates Nrf, which binds to antioxidant response elements (AREs) and induces Phase-I and -II drug-metabolizing enzymes including CYP2B and NQO1. Also increases mRNA levels of multiple hepatic xenobiotic transporters. Elevates expression of genes encoding anti-oxidant and multidrug resistance-associated proteins. Displays chemopreventive activity.
Specifications
Specifications
| Kemisk navn eller materiale | 4-Methyl-5-(2-pyrazinyl)-3H-1,2-dithiole-3-thione |
| CAS | 64224-21-1 |
| Mængde | 10 mg |
| Mål | Nrf2 Modulators |
| Molekylær formel | C8H6N2S3 |
| Formel vægt | 226.34 |
| Renhed | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?