Få mere at vide
Olopatadine Isopropyl Ester, TRC
Artikelnummer. 30428497
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428497 25 mg
30411097 50 mg
30424995 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428497 Leverandør Toronto Research Chemicals Leverandørnr. TRCO57502525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1206456-44-1
Molekylær formel C24 H29 N O3
Molekylvægt (g/mol) 379.49
Synonym (11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid 1-methylethyl ester
IUPAC navn propan-2-yl 2-[(11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetate
SMIL CC(C)OC(=O)Cc1ccc2OCc3ccccc3\C(=C\CCN(C)C)\c2c1
Tekniske data
Kemisk navn eller materiale Olopatadine Isopropyl Ester
Mængde 25 mg
InChI formel InChI=1S/C24H29NO3/c1-17(2)28-24(26)15-18-11-12-23-22(14-18)21(10-7-13-25(3)4)20-9-6-5-8-19(20)16-27-23/h5-6,8-12,14,17H,7,13,15-16H2,1-4H3/b21-10-
Formel vægt 379.2147