Få mere at vide
Olopatadine-d3 N-Oxide, TRC
Artikelnummer. 30425903
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30425903 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425903 Leverandør Toronto Research Chemicals Leverandørnr. TRCO5750121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olopatadine-d3 N-Oxide
CAS 1246832-94-9
Molekylær formel C21H20D3NO4
Mængde 1 mg
Synonym 11-[3-(Dimethyloxidoamino-d3)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic Acid
InChI formel InChI=1S/C21H23NO4/c1-22(2,25)11-5-8-18-17-7-4-3-6-16(17)14-26-20-10-9-15(12-19(18)20)13-21(23)24/h3-4,6-10,12H,5,11,13-14H2,1-2H3,(H,23,24)/b18-8-/i1D3
SMIL [2H]C([2H])([2H])[N+](C)([O-])CC\C=C/1\c2ccccc2COc3ccc(CC(=O)O)cc13
IUPAC navn (3Z)-3-[2-(carboxymethyl)-6H-benzo[c][1]benzoxepin-11-ylidene]-N-methyl-N-(trideuteriomethyl)propan-1-amine oxide
Molekylvægt (g/mol) 356.43
Formel vægt 356.1815