Få mere at vide
Olmesartan Medoxomil-d6, TRC
Artikelnummer. 30418189
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30418189 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418189 Leverandør Toronto Research Chemicals Leverandørnr. TRCO5500031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olmesartan Medoxomil-d6
CAS 1127298-67-2
Molekylær formel C29 H24 D6 N6 O6
Mængde 1 mg
Synonym 4-[1-Hydroxy-1-(methylethyl-d6)]-2-propyl-1-[[2’-(1H-tetazol-5-yl)[1,1’-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl Ester, CS-866-d6, Benicar-d6, Olmetec-d6
InChI formel InChI=1S/C29H30N6O6/c1-5-8-23-30-25(29(3,4)38)24(27(36)39-16-22-17(2)40-28(37)41-22)35(23)15-18-11-13-19(14-12-18)20-9-6-7-10-21(20)26-31-33-34-32-26/h6-7,9-14,38H,5,8,15-16H2,1-4H3,(H,31,32,33,34)/i3D3,4D3
SMIL [2H]C([2H])([2H])C(O)(c1nc(CCC)n(Cc2ccc(cc2)c3ccccc3c4nn[nH]n4)c1C(=O)OCC5=C(C)OC(=O)O5)C([2H])([2H])[2H]
IUPAC navn (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-propyl-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]-5-[2,2,2-trideuterio-1-hydroxy-1-(trideuteriomethyl)ethyl]imidazole-4-carboxylate
Molekylvægt (g/mol) 564.622
Formel vægt 564.2603