Få mere at vide
Olmesartan Acid Ethyl Ester, TRC
Artikelnummer. 30136308
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136308 50 mg
30132899 100 mg
30005150 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136308 Leverandør Toronto Research Chemicals Leverandørnr. TRCO55005050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 144689-23-6
Molekylær formel C26 H30 N6 O3
Molekylvægt (g/mol) 474.55
Synonym 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester, Olmesartan Ethyl Ester
IUPAC navn ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
SMIL CCCc1nc(c(C(=O)OCC)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)C(C)(C)O
Tekniske data
Kemisk navn eller materiale Olmesartan Acid Ethyl Ester
Mængde 50 mg
InChI formel InChI=1S/C26H30N6O3/c1-5-9-21-27-23(26(3,4)34)22(25(33)35-6-2)32(21)16-17-12-14-18(15-13-17)19-10-7-8-11-20(19)24-28-30-31-29-24/h7-8,10-15,34H,5-6,9,16H2,1-4H3,(H,28,29,30,31)
Formel vægt 474.2379