Få mere at vide
Oleuropein Aglycone, TRC
Artikelnummer. 30133055
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133055 100 mg
30138314 50 mg
30005396 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133055 Leverandør Toronto Research Chemicals Leverandørnr. TRCO532945100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31773-95-2
Molekylær formel C19H22O8
Molekylvægt (g/mol) 378.37
Synonym (2R,3E,4S)-3-Ethylidene-3,4-dihydro-2-hydroxy-5-(methoxycarbonyl)-2H-pyran-4-acetic Acid 2-(3,4-Dihydroxyphenyl)ethyl Ester, [2R-(2α,3E,4β)]-3-Ethylidene-3,4-dihydro-2-hydroxy-5-(methoxycarbonyl)-2H-pyran-4-acetic Acid 2-(3,4-Dihydroxyphenyl)ethyl Ester, 5-Carboxy-3-ethylidene-3,4-dihydro-2-hydroxy-2H-pyran-4-acetic Acid 4-(3,4-Dihydroxyphenethyl) 5-Methyl Ester, 3,4-DHPEA-EA, Oleoeuropein Aglycone, Oleoeuropeine Aglycone, Oleuropein Aglycon, Oleuropeine Aglycone
IUPAC navn methyl (4S,5E,6R)-4-[2-[2-(3,4-dihydroxyphenyl)ethoxy]-2-oxoethyl]-5-ethylidene-6-hydroxy-4H-pyran-3-carboxylate
SMIL O=C(OCCC1=CC=C(O)C(O)=C1)C[C@H]2/C([C@H](O)OC=C2C(OC)=O)=C\C
Tekniske data
Kemisk navn eller materiale Oleuropein Aglycone
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C19H22O8/c1-3-12-13(14(18(23)25-2)10-27-19(12)24)9-17(22)26-7-6-11-4-5-15(20)16(21)8-11/h3-5,8,10,13,19-21,24H,6-7,9H2,1-2H3/b12-3+/t13-,19+/m0/s1
Formel vægt 378.1315
Procent renhed >95%