Få mere at vide
STEMCELL Technologies Oleuropein
Beskrivelse
- Oleuropein is the primary phenolic compound found in olives and olive oil, and it exhibits antioxidant, anti-inflammatory, and anti-cancer properties (Sun et al.)
- Decreases adipocyte differentiation and increases osteoblast differentiation in mesenchymal stem cells (Santiago-Mora et al.)
- Inhibits proliferation and induces apoptosis in MCF-7 human breast cancer cells (Han et al.)
Tekniske data
Tekniske data
| CAS | 32619-42-4 |
| Mængde | 25 mg |
| Molekylær formel | C25H32O13 |
| Renhed | ≥98% |