Få mere at vide
Olesoxime, MedChemExpress
Artikelnummer. 16498767
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16498767 5 mg
16496168 10 mg
16472307 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16498767 Leverandør Medchem Express Leverandørnr. HY14796
MedChemExpress Olesoxime (TRO 19622) is a mitochondrial-targeted neuroprotective compound with mean EC50 value for increasing cell survival is 3.2±0.2 μM.
Olesoxime (TRO 19622) is a mitochondrial-targeted neuroprotective compound with mean EC50 value for increasing cell survival is 3.2±0.2 μM.
Tekniske data
Kemisk navn eller materiale Olesoxime
Fysisk form Solid
CAS 22033-87-0
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C27H45NO
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym TRO 19622 NSC 21311
Opløselighedsinformation DMSO : 50 mg/mL (125.11 mM; Need ultrasonic)
SMIL C[C@@]12C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4([C@]([H])(C)CCCC(C)C)[H])=CC(CC1)=NO
Molekylvægt (g/mol) 399.65
Formel vægt 399.65
Procent renhed 98.64%