Få mere at vide
Olaquindox-d4, TRC
Artikelnummer. 30426226
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30426226 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426226 Leverandør Toronto Research Chemicals Leverandørnr. TRCO5150022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Olaquindox-d4
CAS 1189487-82-8
Renhedsgrad noter HPLC
Molekylær formel C12 2H4 H9 N3 O4
Mængde 2.5 mg
Synonym 3-Methyl-1,4-dioxido-N-(1,1,2,2-tetradeuterio-2-hydroxyethyl)quinoxaline-1,4-diium-2-carboxamide
InChI formel InChI=1S/C12H13N3O4/c1-8-11(12(17)13-6-7-16)15(19)10-5-3-2-4-9(10)14(8)18/h2-5,16H,6-7H2,1H3,(H,13,17)/i6D2,7D2
SMIL [2H]C([2H])(O)C([2H])([2H])NC(=O)c1c(C)[n+]([O-])c2ccccc2[n+]1[O-]
IUPAC navn 3-methyl-1,4-dioxido-N-(1,1,2,2-tetradeuterio-2-hydroxyethyl)quinoxaline-1,4-diium-2-carboxamide
Molekylvægt (g/mol) 267.27
Formel vægt 267.1157
Procent renhed >95