Få mere at vide
Olanzapine Pamoate Hydrate, TRC
Artikelnummer. 30417844
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417844 1 g
30410479 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30417844 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2537751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 221373-18-8
Molekylær formel C23 H16 O6 . C17 H20 N4 S . H2 O
Molekylvægt (g/mol) 718.82
Synonym 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 2-methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, hydrate (1:1:1), 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 2-methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine (1:1), monohydrate (9CI), 10H-Thieno[2,3-b][1,5]benzodiazepine, 2-methyl-4-(4-methyl-1-piperazinyl)-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1), monohydrate (9CI), Olanzapine pamoate, Olanzapine pamoate monohydrate, Zyprexa Relprevv, Olanzapine embonate monohydrate, 2-Methyl-4-(4-methylpiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepine 4,4'-methylenebis(3-hydroxynaphthalene-2-carboxylate) monohydrate
IUPAC navn 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;2-methyl-4-(4-methylpiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepine;hydrate
SMIL O.CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24.OC(=O)c5cc6ccccc6c(Cc7c(O)c(cc8ccccc78)C(=O)O)c5O
Tekniske data
Kemisk navn eller materiale Olanzapine Pamoate Hydrate
Mængde 1 g
InChI formel InChI=1S/C23H16O6.C17H20N4S.H2O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12;/h1-10,24-25H,11H2,(H,26,27)(H,28,29);3-6,11,19H,7-10H2,1-2H3;1H2
Formel vægt 718.2461